Vanadium, Plasma Standard Solution, Specpure®, V 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13850-AD
13850-AE
13850-AP
CAAA13850-ADEA
81.05
CAD
CAAA13850-AD
CAAA13850-AE
CAAA13850-AP
Vanadium, Plasma Standard Solution, Specpure®, V 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Vanadium Standards
Molecular Formula: V2O5 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011453
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...