Silver, Plasma Standard Solution, Specpure®, Ag 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13849-AD
13849-AE
13849-AP
CAAA13849-ADEA
86.11
CAD
CAAA13849-AD
CAAA13849-AE
CAAA13849-AP
Silver, Plasma Standard Solution, Specpure®, Ag 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Silver Standards
Molecular Formula: Ag in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00003397
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...