Indium, Plasma Standard Solution, Specpure®, In 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13846-AD
13846-AE
13846-AP
CAAA13846-ADEA
95.12
CAD
CAAA13846-AD
CAAA13846-AE
CAAA13846-AP
Indium, Plasma Standard Solution, Specpure®, In 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Indium Standards
Molecular Formula: In in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134048
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...