Hafnium, plasma standard solution, Specpure®, Hf 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13843-AD
13843-AE
CAAA13843-ADEA
107.12
CAD
CAAA13843-AD
CAAA13843-AE
CAAA13843-AP
Hafnium, plasma standard solution, Specpure®, Hf 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Hafnium Standards
Molecular Formula: HfCl2O in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011032
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...