Specpure® Sulfur, Plasma Standard Solution, S 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13842-AD
13842-AE
13842-AP
CAAA13842-ADEA
85.64
CAD
CAAA13842-AD
CAAA13842-AE
CAAA13842-AP
Specpure® Sulfur, Plasma Standard Solution, S 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Sulfur standards
Molecular Formula: (NH4)2SO4 in H2O
Physical Form: Liquid
MDL No.: MFCD00085316
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...