Germanium, plasma standard solution, Specpure®, Ge 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13841-AD
13841-AE
13841-AP
CAAA13841-ADEA
89.11
CAD
CAAA13841-AD
CAAA13841-AE
CAAA13841-AP
Germanium, plasma standard solution, Specpure®, Ge 1000µg/ml, Thermo Scientific
Standards
Cationic Standards
Germanium Standards
Molecular Formula: Ge in 5% HNO3/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085310
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...