Tantalum, Plasma Standard Solution, Specpure®, Ta 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13840-AD
13840-AE
13840-AP
CAAA13840-ADEA
89.11
CAD
CAAA13840-AD
CAAA13840-AE
CAAA13840-AP
Tantalum, Plasma Standard Solution, Specpure®, Ta 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Tantalum Standards
Molecular Formula: TaCl5 in 2% HF
Physical Form: Liquid
UN No.: 1790
MDL No.: MFCD00011252
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...