Nickel, Plasma Standard Solution, Specpure®, Ni 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13839-AD
13839-AE
13839-AP
CAAA13839-ADEA
85.16
CAD
CAAA13839-AD
CAAA13839-AE
CAAA13839-AP
Nickel, Plasma Standard Solution, Specpure®, Ni 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Nickel Standards
Molecular Formula: Ni in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011137
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...