Dysprosium, plasma standard solution, Specpure®, Dy 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13838-AE
13838-AP
CAAA13838-AEEA
127.35
CAD
CAAA13838-AE
CAAA13838-AP
Dysprosium, plasma standard solution, Specpure®, Dy 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Dysprosium Standards
Molecular Formula: Dy2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010982
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...