Arsenic, plasma standard solution, Specpure®, As 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13836-AD
13836-AE
13836-AP
CAAA13836-ADEA
83.74
CAD
CAAA13836-AD
CAAA13836-AE
CAAA13836-AP
Arsenic, plasma standard solution, Specpure®, As 1000µg/ml, Thermo Scientific
Standards
Cationic Standards
Arsenic Standards
Molecular Formula: As in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085309
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...