Zinc, Plasma Standard Solution, Specpure®, Zn 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13835-AD
13835-AE
13835-AP
CAAA13835-ADEA
83.58
CAD
CAAA13835-AD
CAAA13835-AE
CAAA13835-AP
Zinc, Plasma Standard Solution, Specpure®, Zn 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Zinc Standards
Molecular Formula: Zn in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011291
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...