Palladium, plasma standard solution, Specpure®, Pd 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13833-AD
13833-AE
13833-AP
CAAA13833-ADEA
149.31
CAD
CAAA13833-AD
CAAA13833-AE
CAAA13833-AP
Palladium, plasma standard solution, Specpure®, Pd 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Palladium Standards
Molecular Formula: Pd in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD03613603
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...