Specpure® Niobium, Plasma Standard Solution, Nb 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13831-AD
13831-AE
13831-AP
CAAA13831-ADEA
81.05
CAD
CAAA13831-AD
CAAA13831-AE
CAAA13831-AP
Specpure® Niobium, Plasma Standard Solution, Nb 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Niobium Standards
Molecular Formula: NbCl5 in 2% HF
Physical Form: Liquid
UN No.: 2922
MDL No.: MFCD00011126
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...