Gadolinium, Plasma Standard Solutions, Specpure®, Thermo Scientific

Supplier: Thermo Scientific Chemicals

13829-AD 13829-AE 13829-AP 14373-AD 14373-AE 14373-AP
CAAA13829-ADEA 95.75 CAD
CAAA13829-AD CAAA13829-AE CAAA13829-AP CAAA14373-AD CAAA14373-AE CAAA14373-AP
Gadolinium, Plasma Standard Solutions, Specpure®, Thermo Scientific
Standards Cationic Standards Gadolinium Standards

Molecular Formula: Gd2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011022

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR