Manganese, Plasma Standard Solution, Specpure®, Mn 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13826-AD
13826-AE
13826-AP
CAAA13826-ADEA
85
CAD
CAAA13826-AD
CAAA13826-AE
CAAA13826-AP
Manganese, Plasma Standard Solution, Specpure®, Mn 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Manganese Standards
Molecular Formula: Mn in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011111
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...