Thulium, plasma standard solution, Specpure®, Tm 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13824-AE
13824-AP
CAAA13824-AEEA
161.16
CAD
CAAA13824-AE
CAAA13824-AP
Thulium, plasma standard solution, Specpure®, Tm 1000µg/ml, Thermo Scientific
Standards
Cationic Standards
Thulium Standards
Molecular Formula: Tm2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011281
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...