Lithium, plasma standard solution, Specpure®, Li 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

13821-AD 13821-AE 13821-AP
CAAA13821-ADEA 82.32 CAD
CAAA13821-AD CAAA13821-AE CAAA13821-AP
Lithium, plasma standard solution, Specpure®, Li 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Lithium Standards

Molecular Formula: Li2CO3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134051

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR