Rhenium, plasma standard solution, Specpure®, Re 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

13817-AD 13817-AE 13817-AP
CAAA13817-ADEA 144.73 CAD
CAAA13817-AD CAAA13817-AE CAAA13817-AP
Rhenium, plasma standard solution, Specpure®, Re 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Rhenium Standards

Molecular Formula: Re in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011195

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR