Cerium, plasma standard solution, Specpure®, Ce 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13815-AD
13815-AE
CAAA13815-ADEA
80.26
CAD
CAAA13815-AD
CAAA13815-AE
CAAA13815-AP
Cerium, plasma standard solution, Specpure®, Ce 1000µg/ml, Thermo Scientific
Standards
Cationic Standards
Cerium Standards
Molecular Formula: Ce in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010924
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...