Cadmium Plasma Standard Solutions, for ICP, Specpure®, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13813-AD
13813-AE
13813-AP
14424-AE
14424-AP
CAAA13813-ADEA
83.58
CAD
CAAA13813-AD
CAAA13813-AE
CAAA13813-AP
CAAA14424-AE
CAAA14424-AP
Cadmium Plasma Standard Solutions, for ICP, Specpure®, Thermo Scientific
Standards
Cationic Standards
Cadmium Standards
Molecular Formula: Cd in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010914
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...