Nickel ≥99.8% (metals basis), soluble -325 mesh, Thickness: 0,37 µm
Supplier: Thermo Scientific Chemicals
|
Danger
|
13788-30
13788-A1
CAAA13788-30EA
105.23
CAD
CAAA13788-30
CAAA13788-A1
Nickel ≥99.8% (metals basis), soluble -325 mesh, Thickness: 0,37 µm
Nickel
Purity: 99.8%
CAS No.: 7440-02-0
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00011137
Formula:
Ni Boiling Pt: 3007 °C (1013 hPa) Melting Pt: 1453 °C Density: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1,II Merck Index: 12,06582 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...