Chloropentaamminecobalt(III) chloride
Supplier: Thermo Scientific Chemicals
|
Warning
|
13765-04
13765-09
13765-18
CAAA13765-04EA
93.85
CAD
CAAA13765-04
CAAA13765-09
CAAA13765-18
Chloropentaamminecobalt(III) chloride
Chloropentaamminecobalt (III) chloride
Slightly soluble in water, soluble in conc. H₂SO₄. Insoluble in alcohol
CAS No.: 13859-51-3
Molecular Formula: H15Cl3CoN5
Formula Weight: 250.44
Physical Form: Powder
Density: 1.819
Appearance: Purple
Odor: Odorless
Melting Point: dec.
Solubility: Slightly soluble in water, soluble in conc. H2SO4. Insoluble in alcohol
MDL No.: MFCD00064728
Formula:
H₁₅Cl₃CoN₅ Melting Pt: Dec. Density: 1,819 Storage Temperature: Ambient |
MDL Number:
MFCD00064728 CAS Number: 13859-51-3 EINECS: 237-594-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...