di-Lithium oxalate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dilithium oxalate, Lithium oxalate, Lithiumoxalate
13426-18
13426-30
13426-A1
CAAA13426-18EA
69.2
CAD
CAAA13426-18
CAAA13426-30
CAAA13426-A1
di-Lithium oxalate 99+%
di-Lithium oxalate
Soluble in water
Purity: 99+%
CAS No.: 553-91-3
Molecular Formula: Li2C2O4
Formula Weight: 101.96
Physical Form: Powder
Density: 2.12
Melting Point: dec.
Solubility: Soluble in water
Merck Reference: 14,5537
UN No.: 3288
MDL No.: MFCD00040596
Formula:
C₂Li₂O₄ MW: 101.9 g/mol Melting Pt: Dec. Density: 2.12 Storage Temperature: Ambient |
MDL Number:
MFCD00040596 CAS Number: 553-91-3 EINECS: 209-054-1 UN: 3288 ADR: 6.1 III Merck Index: 14,5537 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...