Europium(III) sulfate octahydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

13210-06 13210-14
CAAA13210-06EA 202.24 CAD
CAAA13210-06 CAAA13210-14
Europium(III) sulfate octahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Europium (III) sulphate octahydrate

Purity: 99.99% CAS No.: 10031-55-7 Molecular Formula: Eu2(SO4)3.8H2O Formula Weight: 736.23 (592.10anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Merck Reference: 14,3904 MDL No.: MFCD00149704

Formula: Eu₂O₁₂S₃·8H₂O
MW: 736.23 g/mol
Melting Pt: 375 °C (-8H₂O)
Storage Temperature: Ambient
MDL Number: MFCD00149703
CAS Number: 10031-55-7
EINECS: 236-903-3
Merck Index: 14,3904

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR