Cadmium perchlorate hexahydrate
Supplier: Thermo Scientific Chemicals
|
Warning
|
12936-18
12936-30
CAAA12936-18EA
115.34
CAD
CAAA12936-18
CAAA12936-30
Cadmium perchlorate hexahydrate
Cadmium perchlorate hexahydrate
CAS No.: 10326-28-0
Molecular Formula: Cd(ClO4)2.6H2O
Formula Weight: 419.39 (311.30anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
MDL No.: MFCD00015992
Formula:
CdCl₂O₈·6H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150202 CAS Number: 10326-28-0 UN: 1481 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...