Bis(cyclopentadienyl)zirconium dichloride

Supplier: Thermo Scientific Chemicals
Warning

12548-06 12548-14 12548-22
CAAA12548-06EA 63.52 CAD
CAAA12548-06 CAAA12548-14 CAAA12548-22
Bis(cyclopentadienyl)zirconium dichloride
Bis(cyclopentadienyl)zirconium dichloride

Fieser: 10,131 15,120 16,121 17,106 18,415 19,414 20,443 21,497


CAS No.: 1291-32-3 Molecular Formula: C10H10Cl2Zr Formula Weight: 292.32 Physical Form: Powder, Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: White to off-white Melting Point: 242-245° Boiling Point: 124-125°/15mm MDL No.: MFCD00003726

Formula: C₁₀H₁₀Cl₂Zr
Boiling Pt: 124…125 °C (15 mmHg)
Melting Pt: 242…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00003726
CAS Number: 1291-32-3
EINECS: 215-066-8
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR