Ammonium nickel (II) sulfate hexahydrate

Supplier: Thermo Scientific Chemicals
Danger

12519-22 12519-36
CAAA12519-22EA 149.78 CAD
CAAA12519-22 CAAA12519-36
Ammonium nickel (II) sulfate hexahydrate
Ammonium nickel (II) sulphate hexahydrate

CAS No.: 7785-20-8 Molecular Formula: H8N2NiO8S2.6H2O Formula Weight: 394.99 (286.90anhy) Physical Form: Large Crystals Density: 1.923 Appearance: Green Odor: Odorless Merck Reference: 14,6501 UN No.: 3077 MDL No.: MFCD00150260

Formula: H₈N₂NiO₈S₂·6H₂O
Density: 1.923
Storage Temperature: Ambient
MDL Number: MFCD00150260
CAS Number: 7785-20-8
EINECS: 239-793-5
UN: 3077
ADR: 9,III
Merck Index: 14,6501

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR