Manganese(II) perchlorate hexahydrate, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

12309-22 12309-36
CAAA12309-22EA 78.84 CAD
CAAA12309-22 CAAA12309-36
Manganese(II) perchlorate hexahydrate, Reagent Grade
Manganese (II) perchlorate hexahydrate

Soluble in benzene, ethyl acetate


CAS No.: 15364-94-0 Molecular Formula: Mn(ClO4)2.6H2O Formula Weight: 361.93 (253.84anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Odor: Odorless UN No.: 1481 MDL No.: MFCD00150255

Formula: Cl₂MnO₈·6H₂O
MW: 361.931 g/mol
Melting Pt: 165 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150255
CAS Number: 15364-94-0
EINECS: 237-390-9
UN: 1481
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR