Ammonium pentaborate octahydrate ≥99.0%
Supplier: Thermo Scientific Chemicals
|
Warning
|
12294-36
12294-A3
CAAA12294-36EA
117.55
CAD
CAAA12294-36
CAAA12294-A3
Ammonium pentaborate octahydrate ≥99.0%
Ammonium pentaborate octahydrate
Purity: 99.0%
CAS No.: 12046-03-6
Molecular Formula: (NH4)2B10O16.8H2O
Formula Weight: 544.29 (400.17anhy)
Physical Form: Powder
Odor: Odorless
MDL No.: MFCD00135839
Formula:
H₄B₅NO₈·8H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00135839 CAS Number: 12046-03-6 EINECS: 234-521-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...