Potassium tetrachloroaurate hydrate ≥99.99% (metals basis) Au ≥ 49%, Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Gold potassium chloride hydrate, Potassium tetrachloroaurate(III) hydrate, Potassium gold(III) chloride hydrate
12150-03
12150-06
CAAA12150-03EA
235.42
CAD
CAAA12150-03
CAAA12150-06
Potassium tetrachloroaurate hydrate ≥99.99% (metals basis) Au ≥ 49%, Premion®
Potassium tetrachloroaurate hydrate
Soluble in water
Purity: 99.99%
CAS No.: 27988-75-6
Molecular Formula: AuCl4K.xH2O
Formula Weight: 377.88(anhy)
Physical Form: Crystalline
Appearance: Yellow to orange
Melting Point: 357° dec.
Solubility: Soluble in water
Merck Reference: 14,7681
UN No.: 3260
MDL No.: MFCD00043088
Formula:
AuCl₄K·H₂O MW: 377.88 g/mol (anhy) Melting Pt: 357 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00043088 CAS Number: 27988-75-6 EINECS: 237-190-1 UN: 3260 ADR: 8 III Merck Index: 14,7681 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...