Hafnium dichloride oxide octahydrate 98+% (metals basis excluding Zr) (max. 1.5% Zr)

Supplier: Thermo Scientific Chemicals
Danger

11833-14 11833-22 11833-36
CAAA11833-14EA 155.95 CAD
CAAA11833-14 CAAA11833-22 CAAA11833-36
Hafnium dichloride oxide octahydrate 98+% (metals basis excluding Zr) (max. 1.5% Zr)
Hafnium dichloride oxide octahydrate

Applications: Synthesis of Hafnium (IV) oxide
Soluble in water


Purity: 98+% CAS No.: 14456-34-9 Molecular Formula: Cl2HfO.8H2O Formula Weight: 409.52 (265.40anhy) Physical Form: Powder Density: 2.2 Appearance: White Odor: Pungent Solubility: Soluble in water UN No.: 3260 MDL No.: MFCD00150417


Synthesis of Hafnium (IV) oxide

Formula: Cl₂HfO·8H₂O
Density: 2.2
Storage Temperature: Ambient
MDL Number: MFCD00150417
CAS Number: 14456-34-9
EINECS: 237-345-3
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR