Silver perchlorate monohydrate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
11624-04
11624-09
11624-18
CAAA11624-04EA
93.06
CAD
CAAA11624-04
CAAA11624-09
CAAA11624-18
CAAA11624-30
Silver perchlorate monohydrate ≥99.9% (metals basis)
Silver perchlorate monohydrate
Fieser: 2,369 7,142 9,413 10,354 11,469 15,121 16,300 18,321 20,341
Purity: 99.9%
CAS No.: 14242-05-8
Molecular Formula: AgClO4.H2O
Formula Weight: 225.34 (207.32anhy)
Physical Form: Crystalline
Density: 2.800
Appearance: White
Odor: Odorless
Melting Point: 485° dec.
Merck Reference: 14,8523
UN No.: 1481
MDL No.: MFCD00149128
Formula:
AgClO₄·1H₂O Melting Pt: 485 °C (dec.) Density: 2.800 Storage Temperature: Ambient |
MDL Number:
MFCD00149128 CAS Number: 14242-05-8 EINECS: 232-035-4 UN: 1481 ADR: 5.1,II Merck Index: 14,8523 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...