Thulium(III) chloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

11201-02 11201-04 11201-09
CAAA11201-02EA 107.28 CAD
CAAA11201-02 CAAA11201-04 CAAA11201-09
Thulium(III) chloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Thulium (III) chloride hydrate

Purity: 99.99% CAS No.: 19423-86-0 Molecular Formula: TmCl3.xH2O Formula Weight: 275.29(anhy) Physical Form: Wet Crystalline Aggregates Sensitivity: Hygroscopic Merck Reference: 14,9394 MDL No.: MFCD00011283

Formula: Cl₃Tm·H₂O
MW: 293.309 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149877
CAS Number: 19423-86-0
EINECS: 236-904-9
Merck Index: 14,9394

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR