Tris(triphenylphosphine)ruthenium(II) chloride ≥99.95% (metals basis) Ru ≥ 10.2%, Premion®
Supplier: Thermo Scientific Chemicals
Synonyms:
Dichlorotris(triphenylphosphine)ruthenium(II), Dichlorotris(triphenylphosphino)ruthenium (II)
Very slightly soluble (with dissociation) in acetone, alcohol, chloroform, ethyl acetate, and toluene
Purity: 99.95%
CAS No.: 15529-49-4
Molecular Formula: RuCl2[P(C6H5)3]3
Formula Weight: 958.86
Physical Form: Crystalline powder
Sensitivity: Air and Moisture Sensitive
Appearance: Brown to black
Melting Point: 132-134°
Solubility: Very slightly soluble (with dissociation) in acetone, alcohol, chloroform, ethyl acetate, and toluene
MDL No.: MFCD00013077
Formula:
C₅₄H₄₅Cl₂P₃Ru MW: 958.85 g/mol Melting Pt: 132…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013077 CAS Number: 15529-49-4 EINECS: 239-569-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...