cis-Dichlorobis(triphenylphosphite)platinum(II) Pt ≥ 24,2%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
cis-Bis(triphenylphosphine)palladium (II) chloride, cis-Bis(triphenylphosphine)platinum(II) chloride, Pt(PPh₃)₂Cl₂
10492-03
10492-MD
CAAA10492-03EA
263.86
CAD
CAAA10492-03
CAAA10492-MD
cis-Dichlorobis(triphenylphosphite)platinum(II) Pt ≥ 24,2%
cis-Dichlorobis(triphenylphosphite)platinum(II)
Slightly soluble in chloroform, hexane toluene
CAS No.: 15604-36-1
Molecular Formula: PtCl2[P(C6H5)3]2
Formula Weight: 790.58
Physical Form: Crystalline
Sensitivity: Hygroscopic
Melting Point: 310° dec.
MDL No.: MFCD00010825
Formula:
[(C₆H₅)₃P]₂PtCl₂ MW: 701.91 Melting Pt: 310 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00010825 CAS Number: 15604-36-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...