trans-Bis(triphenylphosphine)palladium(II) chloride Pd ≥ 14.0%

Supplier: Thermo Scientific Chemicals

Synonyms: trans-Dichlorobis(triphenylphosphine)palladium(II)

10491-03 10491-06 10491-14
CAAA10491-03EA 78.37 CAD
CAAA10491-03 CAAA10491-06 CAAA10491-14
trans-Bis(triphenylphosphine)palladium(II) chloride Pd ≥ 14.0%
trans-Bis(triphenylphosphine)palladium (II) chloride

Applications: Hydrogenation, hydrosilation, carbonylation, oxidation, and C-C bond formation


CAS No.: 13965-03-2 Molecular Formula: C36H30Cl2P2Pd Formula Weight: 701.91 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Yellow Odor: Odorless Melting Point: ca 310° dec. MDL No.: MFCD00009593


Hydrogenation, hydrosilation, carbonylation, oxidation, and C-C bond formation

Formula: C₃₆H₃₀Cl₂P₂Pd
MW: 701.91 g/mol
Melting Pt: ∼ 310 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00009593
CAS Number: 28966-81-6
EINECS: 237-744-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR