Funnel, Powder, Angled, Ace Glass Incorporated
Supplier: Ace Glass
7245-25 7245-15 7245-21 7245-07
71001-676EA 344.17 CAD
71001-676 89083-992 89083-994 89083-996
Funnel, Powder, Angled, Ace Glass Incorporated
Funnels Powder Funnels
Powder funnel angled for use in multi-neck flasks
On flasks with angled necks, this design allows the mouth of the funnel to be positioned back at vertical. Fabricated from heavy wall borosilicate glass.
Order Now
SPECIFICATIONS
Environmentally Preferable