L-Prolinamide 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Pro-NH₂, (S)-2-Pyrrolidinecarboxamide, H-Pro-NH2

L20417-03 L20417-06
CAAAAL20417-03EA 93.38 CAD
CAAAAL20417-03 CAAAAL20417-06
L-Prolinamide 98%
L-Prolinamide

Purity: 99% CAS No.: 7531-52-4 Molecular Formula: C5H10N2O Formula Weight: 114.15 Melting Point: 95-99° Beilstein Reference: 80807 Optical Rotation: -105° (c=2 in ethanol) MDL No.: MFCD00005253

Formula: C₅H₁₀N₂O
MW: 114.15 g/mol
Melting Pt: 95…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00005253
CAS Number: 7531-52-4
EINECS: 231-397-0

Order Now

Specification Test Results

Optical rotation -105° (c=2 in ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR