2,6-Dimethoxy-3-pyridineboronic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20389-03
L20389-06
L20389-MD
CAAAAL20389-03EA
83.42
CAD
CAAAAL20389-03
CAAAAL20389-06
CAAAAL20389-MD
2,6-Dimethoxy-3-pyridineboronic acid 95%
2,6-Dimethoxy-3-pyridineboronic acid
Purity: 95%
CAS No.: 221006-70-8
Molecular Formula: C7H10BNO4
Formula Weight: 182.97
Melting Point: 128-130°
Beilstein Reference: 9391039
Formula:
C₇H₁₀BNO₄ Melting Pt: 128…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788239 CAS Number: 221006-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...