1-Methoxy-2-methyl-2-propanol ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20351-14
L20351-22
CAAAAL20351-14EA
417.12
CAD
CAAAAL20351-14
CAAAAL20351-22
1-Methoxy-2-methyl-2-propanol ≥98%
1-Methoxy-2-methyl-2-propanol
Purity: 98+%
CAS No.: 3587-64-2
Molecular Formula: C5H12O2
Formula Weight: 104.15
Flash Point: 27°(80°F)
Refractive Index: 1.4044
Boiling Point: 115-116°
UN No.: 1987
MDL No.: MFCD03701584
Formula:
C₅H₁₂O₂ Boiling Pt: 115…116 °C Density: 0.892 Flash Pt: 27 °C(80 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03701584 CAS Number: 3587-64-2 UN: 1987 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...