Di-tert-butyl iminodicarboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Di-(tert-butyl) imidodicarbonate

L20325-06 L20325-14
CAAAAL20325-03EA 105.28 CAD
CAAAAL20325-03 CAAAAL20325-06 CAAAAL20325-14
Di-tert-butyl iminodicarboxylate 98%
Di-tert-butyl iminodicarboxylate

Purity: 98% CAS No.: 51779-32-9 Molecular Formula: C10H19NO4 Formula Weight: 217.27 Melting Point: 114-119° Beilstein Reference: 1911172

Formula: C₁₀H₁₉NO₄
MW: 217.27 g/mol
Melting Pt: 114…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00043309
CAS Number: 51779-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR