2-(Trifluoromethyl)phenylglyoxal hydrate 98% (dry weight)
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20280-03
L20280-MD
CAAAAL20280-03EA
230.68
CAD
CAAAAL20280-03
CAAAAL20280-06
CAAAAL20280-MD
2-(Trifluoromethyl)phenylglyoxal hydrate 98% (dry weight)
2-(Trifluoromethyl)phenylglyoxal hydrate
Purity: 98%
CAS No.: 745783-91-9
Molecular Formula: C9H5F3O2.xH2O
Formula Weight: 202.13(anhy)
Melting Point: 36-43°
MDL No.: MFCD05664102
Formula:
C₉H₇F₃O₃ Melting Pt: 36…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD06801323 CAS Number: 745783-91-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...