4-(Trifluoromethyl)phenylglyoxal hydrate 98% (dry weight)
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20275-03
L20275-06
CAAAAL20275-03EA
251.22
CAD
CAAAAL20275-03
CAAAAL20275-06
CAAAAL20275-MD
4-(Trifluoromethyl)phenylglyoxal hydrate 98% (dry weight)
4-(Trifluoromethyl)phenylglyoxal hydrate
Purity: 98%
CAS No.: 1736-56-7
Molecular Formula: C9H5F3O2.xH2O
Formula Weight: 202.13(anhy)
Melting Point: 83-85°
Beilstein Reference: 2261131
MDL No.: MFCD05664098
Formula:
C₉H₅F₃O₂·xH₂O Melting Pt: 83…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD05664098 CAS Number: 1736-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...