3,4-(Methylenedioxy)phenylglyoxal hydrate 97% (dry weight)
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20273-03
L20273-06
CAAAAL20273-03EA
133.19
CAD
CAAAAL20273-03
CAAAAL20273-06
3,4-(Methylenedioxy)phenylglyoxal hydrate 97% (dry weight)
3,4-(Methylenedioxy)phenylglyoxal hydrate
Purity: 97%
CAS No.: 65709-23-1
Molecular Formula: C9H6O4.xH2O
Formula Weight: 178.15(anhy)
Melting Point: 127-131°
Beilstein Reference: 7703971
MDL No.: MFCD06801031
Formula:
C₉H₆O₄·xH₂O Melting Pt: 127…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD06801031 CAS Number: 65709-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...