4-Fluorophenylglyoxal hydrate, hydrate 98% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4-Fluorophenyl)-2-oxoacetaldehyde hydrate, 4-Fluorophenyl(oxo)glyoxal hydrate

L20272-03 L20272-06
CAAAAL20272-03EA 102.38 CAD
CAAAAL20272-03 CAAAAL20272-06
4-Fluorophenylglyoxal hydrate, hydrate 98% (dry weight)
4-Fluorophenylglyoxal hydrate

Purity: 98% CAS No.: 403-32-7 Molecular Formula: C8H5FO2.xH2O Formula Weight: 152.13(anhy) Melting Point: 84-88° Beilstein Reference: 2354778 MDL No.: MFCD01320770

Formula: C₈H₇FO₃
MW: 170.140 g/mol
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD01320770
CAS Number: 7468-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR