3,4-Difluorophenylglyoxal hydrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(3,4-Difluorophenyl)-2-oxoacetaldehyde hydrate
L20271-03
L20271-06
CAAAAL20271-03EA
183.28
CAD
CAAAAL20271-03
CAAAAL20271-06
3,4-Difluorophenylglyoxal hydrate 98%
3,4-Difluorophenylglyoxal hydrate
Purity: 98%
CAS No.: 79784-34-2
Molecular Formula: C8H4F2O2.xH2O
Formula Weight: 170.12(anhy)
Melting Point: 85-90°
Beilstein Reference: 2574671
MDL No.: MFCD03453081
Formula:
C8H6F2O3 MW: 188.13 g/mol Melting Pt: 85…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD03453081 CAS Number: 1049753-77-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...