6-Methoxy-2-naphthylglyoxal hydrate 98% (dry weight)
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20260-03
L20260-06
CAAAAL20260-03EA
113.44
CAD
CAAAAL20260-03
CAAAAL20260-06
6-Methoxy-2-naphthylglyoxal hydrate 98% (dry weight)
6-Methoxy-2-naphthylglyoxal hydrate
Purity: 98%
CAS No.: 1172293-10-5
Molecular Formula: C13H10O3.xH2O
Formula Weight: 214.22(anhy)
Melting Point: 125-130°
MDL No.: MFCD05664091
Formula:
C₁₃H₁₂O₄ Melting Pt: 125…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD05664091 CAS Number: 745783-88-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...