(S)-(+)-1-(1-Naphthyl)ethyl isocyanate 95%

Supplier: Thermo Scientific Chemicals
Warning

L20242-03 L20242-MD
CAAAAL20242-03EA 200.66 CAD
CAAAAL20242-03 CAAAAL20242-MD
(S)-(+)-1-(1-Naphthyl)ethyl isocyanate 95%
(S)-(+)-1-(1-Naphthyl)ethyl isocyanate

Purity: 95% CAS No.: 73671-79-1 Molecular Formula: C13H11NO Formula Weight: 197.24 Storage Temperature: 2°C to 8°C Flash Point: 93°(199°F) Refractive Index: 1.6045 Boiling Point: 106-108°/0.16m Beilstein Reference: 4675069 Optical Rotation: +47° (c=3.5 in toluene) UN No.: 2206 MDL No.: MFCD00064181

Formula: C₁₃H₁₁NO
Boiling Pt: 106…108°/0.16m
Density: 1.118
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064181
CAS Number: 73671-79-1
UN: 2206
ADR: "6.1",III

Order Now

Specification Test Results

Optical rotation +47° (c=3.5 in toluene)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR