(3-Methyl-2-butenyl)tributyltin 95%

Supplier: Thermo Scientific Chemicals
Danger

L20214-03 L20214-06
CAAAAL20214-03EA 113.76 CAD
CAAAAL20214-03 CAAAAL20214-06
(3-Methyl-2-butenyl)tributyltin 95%
(3-Methyl-2-butenyl)tributyltin

Purity: 95% CAS No.: 53911-92-5 Molecular Formula: C17H36Sn Formula Weight: 359.19 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4880 Boiling Point: 283° Beilstein Reference: 3588742 UN No.: 2788 MDL No.: MFCD01863649

Formula: C₁₇H₃₆Sn
Boiling Pt: 283 °C
Density: 1.042
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01863649
CAS Number: 53911-92-5
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR