(1-Propynyl)tributyltin 96%

Supplier: Thermo Scientific Chemicals
Danger

L20201-03 L20201-06
CAAAAL20201-03EA 95.27 CAD
CAAAAL20201-03 CAAAAL20201-06
(1-Propynyl)tributyltin 96%
(1-Propynyl)tributyltin

Purity: 96% CAS No.: 64099-82-7 Molecular Formula: C15H30Sn Formula Weight: 329.12 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4830 Boiling Point: 277° UN No.: 2788 MDL No.: MFCD01863650

Formula: C₁₅H₃₀Sn
Boiling Pt: 277 °C
Density: 1.082
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01863650
CAS Number: 64099-82-7
UN: 2788
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR